Methyl-2-(trifluoromethyl)benzoate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Methyl 2-(trifluoromethyl)benzoate

M2475-5G M2475-25G
TCIAM2475-5GEA 300 NOK
TCIAM2475-5G TCIAM2475-25G
Methyl-2-(trifluoromethyl)benzoate ≥98.0% (by GC)
Methyl-2-(trifluoromethyl)benzoate
Formel: C₉H₇F₃O₂
MW: 204,15 g/mol
Kokepunkt: 105°C
MDL nummer: MFCD00236050
CAS-nummer: 344-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e80eed19e42e5ff3aeb9fbb022b68b6fc73282a&vid=5e80eed19e42e5ff3aeb9fbb022b68b6fc73282a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR