N-(Methoxymethyl)methacrylamide ≥85.0% (by GC) stabilised
Leverandør: TCI
|
Warning
|
Stabilisering:
Stabilised with 4-methoxyphenol (MEHQ)
M2478-25G
M2478-500G
TCIAM2478-25GEA
220
NOK
TCIAM2478-25G
TCIAM2478-500G
N-(Methoxymethyl)methacrylamide ≥85.0% (by GC) stabilised
N-(Methoxymethyl)methacrylamide
Formel:
C₆H₁₁NO₂ MW: 129,16 g/mol Kokepunkt: 85°C |
MDL nummer:
MFCD00035808 CAS-nummer: 3644-12-0 EINECS: 222-872-3 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...