4-Methyl-3-cyclohexene-1-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Methylcyclohex-3-enecarboxylic acid

M2492-1G M2492-5G
TCIAM2492-1GEA 755 NOK
TCIAM2492-1G TCIAM2492-5G
4-Methyl-3-cyclohexene-1-carboxylic acid ≥98.0% (by GC, titration analysis)
4-Methyl-3-cyclohexene-1-carboxylic acid
Formel: C₈H₁₂O₂
MW: 140,18 g/mol
MDL nummer: MFCD02238356
CAS-nummer: 4342-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bab371f159a8f3c91f0d87fdb623f11360423fb&vid=2bab371f159a8f3c91f0d87fdb623f11360423fb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR