1-Methanesulphonyl-2-imidazolidinone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
M2553-5G
M2553-25G
TCIAM2553-5GEA
485
NOK
TCIAM2553-5G
TCIAM2553-25G
1-Methanesulphonyl-2-imidazolidinone ≥98.0% (by GC)
1-Methanesulphonyl-2-imidazolidinone
Formel:
C₄H₈N₂O₃S MW: 164,18 g/mol |
MDL nummer:
MFCD07787278 CAS-nummer: 41730-79-4 EINECS: 255-523-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 191.0 to 195.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...