Metoprolol tartrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

M2555-5G M2555-25G
TCIAM2555-5GEA 770 NOK
TCIAM2555-5G TCIAM2555-25G
Metoprolol tartrate ≥98.0% (by HPLC, titration analysis)
Metoprolol tartrate
Formel: C₁₅H₂₅NO₃
MDL nummer: MFCD00056257
CAS-nummer: 56392-17-7
EINECS: 260-148-9
Merck Indeks: 14,06151
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bdca4b3dd698c331afa4abaeaf4f0273ae0147c&vid=1bdca4b3dd698c331afa4abaeaf4f0273ae0147c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C
Specific rotation [a]20/D +7.0 to +10.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR