3-Metyl-1-buten ∼16,5% ∼2 mol/L in DMF
Leverandør: TCI
|
Danger
|
Synonymer:
Isoamylene, Isopropylethylene, Isopentene
M2564-100ML
M2564-500ML
TCIAM2564-100MLEA
650
NOK
TCIAM2564-100ML
TCIAM2564-500ML
3-Metyl-1-buten ∼16,5% ∼2 mol/L in DMF
3-Metyl-1-buten
Formel:
C₅H₁₀ MW: 70,13 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00008937 CAS-nummer: 563-45-1 EINECS: 209-249-1 FN: 1992 ADR: 3,I |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Concentration (HNMR) | 16.0% to 18.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...