Methyl-3-(4-bromophenyl)propanoate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Methyl 3-(4-Bromophenyl)propionate, 3-(4-Bromophenyl)propionic acid methyll ester, Methyl 3-(4-bromophenyl)propanoate
M2588-1G
M2588-5G
TCIAM2588-1GEA
325
NOK
TCIAM2588-1G
TCIAM2588-5G
Methyl-3-(4-bromophenyl)propanoate ≥98.0% (by GC)
Methyl-3-(4-bromophenyl)propanoate
Formel:
C₁₀H₁₁BrO₂ MW: 243,10 g/mol Kokepunkt: 92°C |
MDL nummer:
MFCD09953150 CAS-nummer: 75567-84-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...