2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Methylthiazole-5-carboxylic acid

M2626-1G M2626-5G
TCIAM2626-1GEA 1050 NOK
TCIAM2626-1G TCIAM2626-5G
2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-5-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
MW: 143,1637 g/mol
MDL nummer: MFCD01760300
CAS-nummer: 40004-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be7aecc0549982a5246d319bd4f4c586eb356206&vid=be7aecc0549982a5246d319bd4f4c586eb356206&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR