Methoxymethyl phenyl sulphone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
M2629-1G
M2629-5G
TCIAM2629-1GEA
675
NOK
TCIAM2629-1G
TCIAM2629-5G
Methoxymethyl phenyl sulphone ≥98.0% (by GC)
Methoxymethyl phenyl sulphone
Formel:
C₈H₁₀O₃S MW: 186,23 g/mol |
CAS-nummer:
15251-78-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...