Methyl-3-methylthiophene-2-carboxylate ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
Methyl 3-methylthiophene-2-carboxylate
M2662-1G
M2662-5G
TCIAM2662-1GEA
380
NOK
TCIAM2662-1G
TCIAM2662-5G
Methyl-3-methylthiophene-2-carboxylate ≥97.0% (by GC)
Methyl-3-methylthiophene-2-carboxylate
Formel:
C₇H₈O₂S MW: 156,20 g/mol Kokepunkt: 217°C |
MDL nummer:
MFCD00234332 CAS-nummer: 81452-54-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...