Mal-amido-PEG4-acid ≥95.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic acid, 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic acid

M2666-25MG M2666-100MG
TCIAM2666-25MGEA 365 NOK
TCIAM2666-25MG TCIAM2666-100MG
Mal-amido-PEG4-acid ≥95.0% (by HPLC)
Mal-amido-PEG4-acid
Formel: C₁₈H₂₈N₂O₉
MW: 416,42 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD13184954
CAS-nummer: 1263045-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be8ed87ceecf2953c8be67cb7e48e8ac42d9e212&vid=be8ed87ceecf2953c8be67cb7e48e8ac42d9e212&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 6.10 to 7.40 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR