(6-Methyl-3-pyridinyl)methanol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline

M2683-1G M2683-5G
TCIAM2683-1GEA 420 NOK
TCIAM2683-1G TCIAM2683-5G
(6-Methyl-3-pyridinyl)methanol ≥97.0% (by GC, titration analysis)
(6-Methyl-3-pyridinyl)methanol
Formel: C₇H₉NO
MW: 123,16 g/mol
Kokepunkt: 100°C
MDL nummer: MFCD08692003
CAS-nummer: 34107-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17ac23a9b89b4f31b97c52de404afcb3a0baa079&vid=17ac23a9b89b4f31b97c52de404afcb3a0baa079&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR