3,5-Dimethyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 4-Methoxy-3,5-dimethylphenylboronic acid, 4-Methoxy-3,5-dimethylbenzeneboronic acid

M2685-1G M2685-5G
TCIAM2685-1GEA 1050 NOK
TCIAM2685-1G TCIAM2685-5G
3,5-Dimethyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethyl-4-methoxyphenylboronic acid
Formel: C₉H₁₃BO₃
MW: 180,01 g/mol
Smeltepunkt: 240°C
MDL nummer: MFCD01114648
CAS-nummer: 301699-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d91dc23de3bd0973627545fd8efa3a65f12a2d76&vid=d91dc23de3bd0973627545fd8efa3a65f12a2d76&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR