5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 5-Methyl-1-phenylpyrazole-4-carboxylic acid

M2692-1G M2692-5G
TCIAM2692-1GEA 660 NOK
TCIAM2692-1G TCIAM2692-5G
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₁H₁₀N₂O₂
MW: 202,21 g/mol
MDL nummer: MFCD00067831
CAS-nummer: 91138-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b75748c70f39894cc518122b35accc93480a69c&vid=5b75748c70f39894cc518122b35accc93480a69c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR