Methyl-6-(hydroxymethyl)nicotinate ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 6-(Hydroxymethyl)pyridine-3-carboxylate methyll ester, Methyl 6-(hydroxymethyl)nicotinate, 6-(Hydroxymethyl)nicotinic acid methyll ester, Methyl 6-(Hydroxymethyl)pyridine-3-carboxylate

M2750-200MG M2750-1G
TCIAM2750-200MGEA 460 NOK
TCIAM2750-200MG TCIAM2750-1G
Methyl-6-(hydroxymethyl)nicotinate ≥97.0% (by GC, titration analysis)
Methyl-6-(hydroxymethyl)nicotinate
Formel: C₈H₉NO₃
MW: 167,1600 g/mol
MDL nummer: MFCD00643821
CAS-nummer: 56026-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a23f71dd00c18624bc2bd6606bebb8bd2202463f&vid=a23f71dd00c18624bc2bd6606bebb8bd2202463f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR