N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulphonamide ≥98.0% (by GC, total nitrogen)

Leverandør: TCI

M2782-1G M2782-5G
TCIAM2782-1GEA 2350 NOK
TCIAM2782-1G TCIAM2782-5G
N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulphonamide ≥98.0% (by GC, total nitrogen)
N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulphonamide
Formel: C₉H₁₀F₃NO₂S₂
MW: 285,30 g/mol
MDL nummer: MFCD29763020
CAS-nummer: 1045822-31-8

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR