2-Methyl-4-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2-Methylthiazole-4-carboxylic acid
M2802-1G M2802-5G
TCIAM2802-1GEA 890 NOK
TCIAM2802-1G TCIAM2802-5G
2-Methyl-4-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-4-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
MW: 143,1637 g/mol
Smeltepunkt: 144°C
MDL nummer: MFCD03407332
CAS-nummer: 35272-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb760ef172266a2b957a5bb695ddcad26c98eea&vid=4bb760ef172266a2b957a5bb695ddcad26c98eea&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %