4-Methoxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 4-Methoxy-3-methylbenzeneboronic acid

M2809-1G M2809-5G
TCIAM2809-1GEA 380 NOK
TCIAM2809-1G TCIAM2809-5G
4-Methoxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Methoxy-3-methylphenylboronic acid
Formel: C₈H₁₁BO₃
MW: 165,99 g/mol
MDL nummer: MFCD02179464
CAS-nummer: 175883-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16facab24d87c3adfc32c074ab3d35993c141548&vid=16facab24d87c3adfc32c074ab3d35993c141548&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR