2-Methyl-9,10-di(2-naphthyl)anthracene ≥97.0% (by HPLC)
Leverandør: TCI
M2821-200MG
M2821-1G
TCIAM2821-200MGEA
1150
NOK
TCIAM2821-200MG
TCIAM2821-1G
2-Methyl-9,10-di(2-naphthyl)anthracene ≥97.0% (by HPLC)
2-Methyl-9,10-di(2-naphthyl)anthracene
Formel:
C₃₅H₂₄ MW: 444,58 g/mol |
MDL nummer:
MFCD12022447 CAS-nummer: 804560-00-7 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...