(1-Methyl-1H-pyrazol-5-yl)methanol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 5-(Hydroxymethyl)-1-methylpyrazole, 1-Methylpyrazole-5-methanol
M2831-200MG M2831-1G
TCIAM2831-200MGEA 350 NOK
TCIAM2831-200MG TCIAM2831-1G
(1-Methyl-1H-pyrazol-5-yl)methanol ≥98.0% (by GC, titration analysis)
(1-Methyl-1H-pyrazol-5-yl)methanol
Formel: C₅H₈N₂O
MW: 112,13 g/mol
MDL nummer: MFCD08556301
CAS-nummer: 84547-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95eb73fd84d33242d27ae9ddd4d16a738a25a3c9&vid=95eb73fd84d33242d27ae9ddd4d16a738a25a3c9&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 56.0 to 60.0 deg-C