4-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: (4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid

M2840-1G M2840-5G
TCIAM2840-1GEA 595 NOK
TCIAM2840-1G TCIAM2840-5G
4-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
4-Methyl-2-thiopheneboronic acid
Formel: C₅H₇BO₂S
MW: 141,99 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01319040
CAS-nummer: 162607-15-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=569a75159d895b73faf634afa411a7354a9b4e98&vid=569a75159d895b73faf634afa411a7354a9b4e98&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR