Methyl-4-chloro-4-oxobutanoate ≥95.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Methyl succinyl chloride, Succinic acid monomethyl ester chloride, Methyl 4-Chloro-4-oxobutyrate, Methyl Succinyl chloride, 4-Chloro-4-oxobutyric acid methyll ester
M2850-25G
M2850-5G
TCIAM2850-25GEA
1800
NOK
TCIAM2850-25G
TCIAM2850-5G
Methyl-4-chloro-4-oxobutanoate ≥95.0% (by GC)
Methyl-4-chloro-4-oxobutanoate
Formel:
C₅H₇ClO₃ MW: 150,56 g/mol Kokepunkt: 65°C Flash punkt: 74°C |
MDL nummer:
MFCD00000750 CAS-nummer: 1490-25-1 EINECS: 216-077-0 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...