Magnesiumbis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 3-nitro-1H-pyrimidine-2,4-dione, Magnesium(II) triflimide, Bis(trifluoromethanesulphonyl)imide Magnesium(II) salt, 2,4(1H,3H)-Pyrimidinedione, nitro-, Magnesium bis(trifluoromethylsulphonyl)imide, Magnesium(II) Bis(trifluoromethanesulphonyl)imide, 3-Nitropyrimidine-2,4(1H,3H)-dione

M2861-1G M2861-5G
TCIAM2861-1GEA 580 NOK
TCIAM2861-1G TCIAM2861-5G
Magnesiumbis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)
Magnesiumbis(trifluoromethylsulphonyl)imide
Formel: C₄F₁₂MgN₂O₈S₄
MW: 584,6 g/mol
MDL nummer: MFCD14156020
CAS-nummer: 133395-16-1
FN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af4df9c6112b31fd2244563debeba0a3a1251b6f&vid=af4df9c6112b31fd2244563debeba0a3a1251b6f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR