Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: HEHEHP, 2-Ethylhexyl Hydrogen (2-Ethylhexyl)phosphonate, (2-Ethylhexyl)phosphonic acid Mono-2-ethylhexyl ester, PC 88A

M2871-5ML M2871-25ML M2871-100ML
TCIAM2871-5MLEA 850 NOK
TCIAM2871-5ML TCIAM2871-25ML TCIAM2871-100ML
Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate ≥95.0% (by titrimetric analysis)
Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate
Formel: C₁₆H₃₅O₃P
MW: 306,43 g/mol
Flash punkt: 173°C
MDL nummer: MFCD00467188
CAS-nummer: 14802-03-0
EINECS: 238-865-3
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa03f00d7e229071c7c125f15327641530ef01e&vid=6fa03f00d7e229071c7c125f15327641530ef01e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR