Methyl-4-methoxy-3-nitrobenzoate ≥98.0% (by GC)
Leverandør: TCI
Synonymer: 4-Methoxy-3-nitrobenzoic acid methyl ester, Methyl 4-methoxy-3-nitrobenzoate
M2873-1G M2873-5G
TCIAM2873-1GEA 445 NOK
TCIAM2873-1G TCIAM2873-5G
Methyl-4-methoxy-3-nitrobenzoate ≥98.0% (by GC)
Methyl-4-methoxy-3-nitrobenzoate
Formel: C₉H₉NO₅
MW: 211,17 g/mol
MDL nummer: MFCD00024213
CAS-nummer: 40757-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163ddd696b93d7656942df2380fbc75e9bab6cdf&vid=163ddd696b93d7656942df2380fbc75e9bab6cdf&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C