methyl cyclobutanecarboxylate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Methyl cyclobutanecarboxylate

M2884-5G M2884-25G
TCIAM2884-5GEA 650 NOK
TCIAM2884-5G TCIAM2884-25G
methyl cyclobutanecarboxylate ≥98.0% (by GC)
methyl cyclobutanecarboxylate
Formel: C₆H₁₀O₂
MW: 114,144 g/mol
Kokepunkt: 135°C
MDL nummer: MFCD00095142
CAS-nummer: 765-85-5
FN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df9c444d870c7eb7ad0be5fd27cb487a8b46fea7&vid=7ddceb97c611f4601522ccb979413860f0b458db&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR