4-Methylumbelliferyl beta-D-Glucuronide hydrate ≥98.0% (by HPLC)
Leverandør: TCI
M3026-100MG
M3026-1G
TCIAM3026-100MGEA
700
NOK
TCIAM3026-100MG
TCIAM3026-1G
4-Methylumbelliferyl beta-D-Glucuronide hydrate ≥98.0% (by HPLC)
4-Methylumbelliferyl beta-D-Glucuronide hydrate
Formel:
C₁₆H₁₆O₉·xH₂O MW: 352,30 g/mol |
MDL nummer:
MFCD09039280 CAS-nummer: 881005-91-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -115 to -120 deg (C=0.2, H2O) (calcd.on anh.substance) |
Water | 5.0 to 17.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...