Methyl-PEG24-Amine ≥98.0% (by HPLC)
Leverandør: TCI
|
Warning
|
Synonymer:
m-PEG24-NH2, 2,5,8,11,14,17,20,23,26,29,32,35,38,41,44,47,50,53,56,59,62,65,68,71-Tetracosaoxatriheptacontan-73-amine, m-PEG24-Amine
M3048-100MG
M3048-25MG
TCIAM3048-100MGEA
6125
NOK
TCIAM3048-100MG
TCIAM3048-25MG
Methyl-PEG24-Amine ≥98.0% (by HPLC)
Methyl-PEG24-Amine
Formel:
C₄₉H₁₀₁NO₂₄ MW: 1088,33 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD11041163 CAS-nummer: 2151823-08-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Elemental analysis (Carbon) | 53.00 to 55.00 % |
Elemental analysis (Nitrogen) | 1.00 to 1.60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...