Methyl p-tolyl sulphoxide ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1-Methyl-4-(methylsulphinyl)benzene, Methyl 4-Tolyl sulphoxide

M3061-1G M3061-5G
TCIAM3061-1GEA 1075 NOK
TCIAM3061-1G TCIAM3061-5G
Methyl p-tolyl sulphoxide ≥98.0% (by GC)
Methyl p-tolyl sulphoxide
Formel: C₈H₁₀OS
MW: 154,227 g/mol
Kokepunkt: 114°C
MDL nummer: MFCD00075175
CAS-nummer: 934-72-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR