6-Methyl-1,3-benzoxazole-2(3H)-thione ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 6-Methyl-3H-benzooxazole-2-thione

M3063-1G M3063-5G
TCIAM3063-1GEA 1125 NOK
TCIAM3063-1G TCIAM3063-5G
6-Methyl-1,3-benzoxazole-2(3H)-thione ≥98.0% (by GC, titration analysis)
6-Methyl-1,3-benzoxazole-2(3H)-thione
Formel: C₈H₇NOS
MW: 165,21 g/mol
MDL nummer: MFCD00175644
CAS-nummer: 23417-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4f0192636304753f88a89869a1cec3a8edbb7a9&vid=ec57ca819bf3cafd246a7e998a740a33fd43a384&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR