2-Methyl-N-(m-tolyl)aniline ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Methyl-N-(3-methylphenyl)benzenamine, 2,3'-Dimethyldiphenylamine, 2-Methyl-N-(3-methylphenyl)aniline
M3064-1G
M3064-5G
TCIAM3064-1GEA
1300
NOK
TCIAM3064-1G
TCIAM3064-5G
2-Methyl-N-(m-tolyl)aniline ≥98.0% (by GC)
2-Methyl-N-(m-tolyl)aniline
Formel:
C₁₄H₁₅N MW: 197,28 g/mol Kokepunkt: 187°C |
CAS-nummer:
34801-11-1 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...