4-(4-Methoxyphenyl)thieno[2,3-b:5,4-c']dipyridine-2-carboxamide ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
TH
M3085-10MG
M3085-50MG
TCIAM3085-10MGEA
2200
NOK
TCIAM3085-10MG
TCIAM3085-50MG
4-(4-Methoxyphenyl)thieno[2,3-b:5,4-c']dipyridine-2-carboxamide ≥98.0% (by HPLC)
4-(4-Methoxyphenyl)thieno[2,3-b:5,4-c']dipyridine-2-carboxamide
Formel:
C₁₈H₁₃N₃O₂S MW: 335,38 g/mol |
CAS-nummer:
942135-14-0 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 11.90 to 13.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...