N-Methoxy-N-methylnicotinamide ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
N-methoxy-N-methylnicotinamide
M3099-1G
M3099-5G
TCIAM3099-1GEA
890
NOK
TCIAM3099-1G
TCIAM3099-5G
N-Methoxy-N-methylnicotinamide ≥96.0% (by GC)
N-Methoxy-N-methylnicotinamide
Formel:
C₈H₁₀N₂O₂ MW: 166,18 g/mol |
MDL nummer:
MFCD11847417 CAS-nummer: 95091-91-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...