7-Methoxy-2,3-dihydro-1H-inden-1-one ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 7-Methoxy-1-indanone, 7-Methoxy-2,3-dihydro-1H-inden-1-one

M3126-1G M3126-200MG
TCIAM3126-1GEA 2225 NOK
TCIAM3126-1G TCIAM3126-200MG
7-Methoxy-2,3-dihydro-1H-inden-1-one ≥98.0% (by GC)
7-Methoxy-2,3-dihydro-1H-inden-1-one
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokepunkt: 110°C
MDL nummer: MFCD07784339
CAS-nummer: 34985-41-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR