2-Methyladamantan-2-yl acrylate ≥98.0% (by GC) stabilised
Leverandør: TCI
|
Warning
|
Synonymer:
Acrylic acid 2-Methyladamantan-2-yl ester
Stabilisering:
Stabilised with 4-methoxyphenol (MEHQ)
M3223-5G
M3223-25G
TCIAM3223-5GEA
865
NOK
TCIAM3223-5G
TCIAM3223-25G
2-Methyladamantan-2-yl acrylate ≥98.0% (by GC) stabilised
2-Methyladamantan-2-yl acrylate
Formel:
C₁₄H₂₀O₂ MW: 220,31 g/mol |
MDL nummer:
MFCD08704223 CAS-nummer: 249562-06-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...