1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-[1-(2-Methoxyethyl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid Pinacol ester, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester

M3232-200MG M3232-1G
TCIAM3232-200MGEA 310 NOK
TCIAM3232-200MG TCIAM3232-1G
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₂H₂₁BN₂O₃
MW: 252,1177 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD12405649
CAS-nummer: 847818-71-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR