4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
2-[(4-MethoxyCarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid methyll ester, Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, Methyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 4-(MethoxyCarbonylmethyl)phenylboronic acid Pinacol ester
M3236-1G
M3236-5G
TCIAM3236-1GEA
515
NOK
TCIAM3236-1G
TCIAM3236-5G
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester
Formel:
C₁₅H₂₁BO₄ MW: 276,1358 g/mol |
MDL nummer:
MFCD09997797 CAS-nummer: 454185-98-9 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...