3-Methoxy-N,N-dimethylpropanamide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Methoxy-N,N-dimethyl-propionamide

M3253-100G M3253-25G
TCIAM3253-100GEA 1550 NOK
TCIAM3253-100G TCIAM3253-25G
3-Methoxy-N,N-dimethylpropanamide ≥98.0% (by GC)
3-Methoxy-N,N-dimethylpropanamide
Formel: C₆H₁₃NO₂
MW: 131,18 g/mol
Kokepunkt: 58°C
MDL nummer: MFCD18970488
CAS-nummer: 53185-52-7
EINECS: 258-420-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR