Methyl-2-(1H-indol-3-yl)-2-oxoacetate ≥96.0% (by GC)
Leverandør: TCI
Synonymer:
Methyl 3-Indoleglyoxylate, Methyl indole-3-glyoxylate, Methyl 2-(1H-indol-3-yl)-2-oxoacetate, 2-(1H-Indol-3-yl)-2-oxoacetic acid methyll ester, 3-Indoleglyoxylic acid methyll ester, Methyl 2-(1H-Indol-3-yl)-2-oxoacetate
M3254-1G
M3254-5G
TCIAM3254-1GEA
435
NOK
TCIAM3254-1G
TCIAM3254-5G
Methyl-2-(1H-indol-3-yl)-2-oxoacetate ≥96.0% (by GC)
Methyl-2-(1H-indol-3-yl)-2-oxoacetate
Formel:
C₁₁H₉NO₃ MW: 203,1941 g/mol |
MDL nummer:
MFCD00047173 CAS-nummer: 18372-22-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 228.0 to 232.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...