7-Methyl-1-tetralone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
7-Methyl-1,2,3,4-tetrahydronaphthalen-1-one, 7-Methyl-3,4-dihydronaphthalen-1(2H)-one
M3255-1G
M3255-5G
TCIAM3255-1GEA
555
NOK
TCIAM3255-1G
TCIAM3255-5G
7-Methyl-1-tetralone ≥98.0% (by GC)
7-Methyl-1-tetralone
Formel:
C₁₁H₁₂O MW: 160,22 g/mol Kokepunkt: 144°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00087600 CAS-nummer: 22009-37-6 EINECS: 244-717-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...