Morpholine hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: Morpholinium chloride
M3284-25G M3284-5G
TCIAM3284-25GEA 2200 NOK
TCIAM3284-25G TCIAM3284-5G
Morpholine hydrochloride ≥98.0% (by titrimetric analysis)
Morpholine hydrochloride
Formel: C₄H₁₀ClNO
MW: 123,58 g/mol
MDL nummer: MFCD00035287
CAS-nummer: 10024-89-2
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 177.0 to 181.0 deg-C