2-Methoxy-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
Leverandør: TCI
M3301-1G
M3301-5G
TCIAM3301-1GEA
475
NOK
TCIAM3301-1G
TCIAM3301-5G
2-Methoxy-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
2-Methoxy-9,9-dimethyl-9H-fluorene
Formel:
C₁₆H₁₆O MW: 224,30 g/mol |
MDL nummer:
MFCD32263492 CAS-nummer: 1514864-84-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...