Nitron ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 1,4-Diphenyl-endoanilino-dihydrotriazole

N0211-25G N0211-5G
TCIAN0211-25GEA 1625 NOK
TCIAN0211-25G TCIAN0211-5G
Nitron ≥98.0% (by titrimetric analysis)
Nitron
Formel: C₂₀H₁₆N₄
MW: 312,38 g/mol
MDL nummer: MFCD00005174
CAS-nummer: 2218-94-2
EINECS: 218-724-2
Merck Indeks: 14,06612

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetic acid almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR