4-(4-Nitrophenylazo)orcinol ≥80.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 4-Nitrobenzene-azo-orcinol, 4-(4-Nitrophenylazo)-5-methylresorcinol

N0324-1G N0324-5G
TCIAN0324-1GEA 1450 NOK
TCIAN0324-1G TCIAN0324-5G
4-(4-Nitrophenylazo)orcinol ≥80.0% (by HPLC)
4-(4-Nitrophenylazo)orcinol
Formel: C₁₃H₁₁N₃O₄
MW: 273,25 g/mol
MDL nummer: MFCD00014710
CAS-nummer: 607-96-5
EINECS: 210-150-0

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 80.0 area%
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR