γ-Linolenic acid
Leverandør: Cayman Chemical
Synonymer: cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid
90220-100 90220-250 90220-1
CAYM90220-100EA 1200 NOK
CAYM90220-100 CAYM90220-250 CAYM90220-1
γ-Linolenic acid
γ-Linolenic acid
Formel: C₁₈H₃₀O₂
MW: 278,44 g/mol
MDL nummer: MFCD00065718
CAS-nummer: 506-26-3