2-Nitrobenzaldehyde oxime ≥85.0% (by GC) deprotecting agent

Leverandør: TCI
Warning

Synonymer: 2-Nitrobenzaldoxime

N0486-25G N0486-5G
TCIAN0486-25GEA 945 NOK
TCIAN0486-25G TCIAN0486-5G
2-Nitrobenzaldehyde oxime ≥85.0% (by GC) deprotecting agent
2-Nitrobenzaldehyde oxime
Formel: C₇H₆N₂O₃
MDL nummer: MFCD00007187
CAS-nummer: 6635-41-2
EINECS: 229-634-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bcf172131e3118aebac85738fd2858e5582a636&vid=3bcf172131e3118aebac85738fd2858e5582a636&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 85.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR