5-Norbornene-2-carboxaldehyde ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde
N0504-25ML
N0504-5ML
TCIAN0504-25MLEA
1175
NOK
TCIAN0504-25ML
TCIAN0504-5ML
5-Norbornene-2-carboxaldehyde ≥95.0% (by GC)
5-Norbornene-2-carboxaldehyde
Formel:
C₈H₁₀O MW: 122,17 g/mol Kokepunkt: 72°C Flash punkt: 50°C |
MDL nummer:
MFCD00167568 CAS-nummer: 5453-80-5 EINECS: 226-698-9 FN: 1989 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0% (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...