2,3-Naphthalenedicarboxylic anhydride ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Naphtho[2,3-c]furan-1,3-dione

N0530-25G N0530-250G
TCIAN0530-25GEA 1175 NOK
TCIAN0530-25G TCIAN0530-250G
2,3-Naphthalenedicarboxylic anhydride ≥95.0% (by GC, titration analysis)
2,3-Naphthalenedicarboxylic anhydride
Formel: C₁₂H₆O₃
MW: 198.18 g/mol
MDL nummer: MFCD00059091
CAS-nummer: 716-39-2
EINECS: 211-936-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Methanolysis Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR