3-(5-Nitro-2-furyl)acrylic acid ≥97.0% (by HPLC)
Leverandør: TCI
|
Warning
|
N0551-25G
N0551-5G
TCIAN0551-25GEA
5300
NOK
TCIAN0551-25G
TCIAN0551-5G
3-(5-Nitro-2-furyl)acrylic acid ≥97.0% (by HPLC)
3-(5-Nitro-2-furyl)acrylic acid
Formel:
C₇H₅NO₅ MW: 183,12 g/mol |
MDL nummer:
MFCD00143078 CAS-nummer: 6281-23-8 EINECS: 228-488-2 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...