N-1-Naphthylethylenediamine dihydrochloride monomethanolate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Marshall's Reagent, Bratton-Marshall Reagent

N0577-1G N0577-25G
TCIAN0577-1GEA 230 NOK
TCIAN0577-1G TCIAN0577-25G
N-1-Naphthylethylenediamine dihydrochloride monomethanolate ≥97.0% (by titrimetric analysis)
N-1-Naphthylethylenediamine dihydrochloride monomethanolate
Formel: C₁₂H₁₄N₂
MW: 291,22 g/mol
Smeltepunkt: 190°C
MDL nummer: MFCD00012556
EINECS: 215-981-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2beff6d975283f3c6446d4658fc27c9c7b4ba972&vid=67139b4dee023fa7ad6472c1b063bb2ad4107bfc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in Water almost transparency
Methanol 8.0 to 11.0 %
Sensitiveness Colorimetric Test : Turn Red with 0.02% Sulfanilic Acid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR